CAS RN 917566-86-0
IUPAC Name:
2-bromo-4-(furan-2-yl)-1,3-thiazole
Synonyms:
2-bromo-4-(furan-2-yl)-1,3-thiazole
Molecular Weight (g/mol):
230.08
Molecular Formula:
C7H4BrNOS
InChi Key:
PUFWLSRDFTVPMS-UHFFFAOYSA-N
SMILES:
BrC1=NC(=CS1)C1=CC=CO1
1
–
1
of
1
results
1
2-Bromo-4-(2-furyl)thiazole, 97%, Thermo Scientific™
CAS: 917566-86-0 Molecular Formula: C7H4BrNOS Molecular Weight (g/mol): 230.079 MDL Number: MFCD11220134 InChI Key: PUFWLSRDFTVPMS-UHFFFAOYSA-N Synonym: 2-bromo-4-furan-2-yl-1,3-thiazole,2-bromo-4-2-furyl thiazole,2-bromo-4-2-furanyl thiazole,2-bromo-4-furan-2-yl thiazole,thiazole, 2-bromo-4-2-furanyl PubChem CID: 38996209 IUPAC Name: 2-bromo-4-(furan-2-yl)-1,3-thiazole SMILES: C1=COC(=C1)C2=CSC(=N2)Br