accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91796-57-5

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexyl 4-methylbenzene-1-sulfinate
Synonyms: 2-isopropyl-5-methylcyclohexyl 4-methylbenzenesulfinate
Molecular Weight (g/mol): 294.45
Molecular Formula: C17H26O2S
InChi Key: NQICGNSARVCSGJ-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OS(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results