accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 918812-18-7

ONSOONH2HCl

CAS RN 918812-18-7

IUPAC Name: 1-[2-(morpholine-4-sulfonyl)phenyl]methanamine hydrochloride
Synonyms: 1-[2-(morpholine-4-sulfonyl)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 292.78
Molecular Formula: C11H17ClN2O3S
InChi Key: RMMDNVBLYNZQFP-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=CC=C1S(=O)(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results