accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NNONH3CH2OCl

CAS RN 919102-70-8

IUPAC Name: 4-(2,4,6-trimethylphenyl)-8-oxa-2λ⁵,4,5-triazatetracyclo[7.7.0.0²,⁶.0¹¹,¹⁶]hexadeca-2,5,11,13,15-pentaen-2-ylium hydrate chloride
Synonyms: 4-(2,4,6-trimethylphenyl)-8-oxa-2λ⁵,4,5-triazatetracyclo[7.7.0.0²,⁶.0¹¹,¹⁶]hexadeca-2,5,11,13,15-pentaen-2-ylium hydrate chloride
Molecular Weight (g/mol): 385.89
Molecular Formula: C21H24ClN3O2
InChi Key: DFFDIRURHBNRTR-UHFFFAOYNA-M
SMILES: O.[Cl-].CC1=CC(C)=C(N2C=[N+]3C4C(CC5=CC=CC=C45)OCC3=N2)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results