accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91976-98-6

IUPAC Name: 4-chloro-N,N-dipropylbutanamide
Synonyms: 4-chloro-N,N-dipropylbutanamide
Molecular Weight (g/mol): 205.73
Molecular Formula: C10H20ClNO
InChi Key: QMFTZGZZBHNKJU-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results