accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 91978-69-7

IUPAC Name: potassium 4-methylphenyl sulfate
Synonyms: potassium 4-methylphenyl sulfate
Molecular Weight (g/mol): 226.29
Molecular Formula: C7H7KO4S
InChi Key: HTSFIPMTBJHYFQ-UHFFFAOYSA-M
SMILES: [K+].CC1=CC=C(OS([O-])(=O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results