accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-15-9

IUPAC Name: N-(2-methoxyphenyl)-3-oxobutanamide
Synonyms: acetoacet-o-anisidide
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: KYYRTDXOHQYZPO-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1NC(=O)CC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results