accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-44-4

IUPAC Name: naphthalene-2,3-diol
Synonyms: 2,3-dihydroxynaphthalene 2,3-dihydroxynaphthlene 2,3-dihydroxynapthalene
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: JRNGUTKWMSBIBF-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (4)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)