accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-53-5

IUPAC Name: 4-phenylmorpholine
Synonyms: N-phenylmorpholine
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: FHQRDEDZJIFJAL-UHFFFAOYSA-N
SMILES: C1CN(CCO1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results