accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-54-6

IUPAC Name: hydrogen 1-phenylpiperazine chloride
Synonyms: phenylpiperazine
Molecular Weight (g/mol): 198.69
Molecular Formula: C10H15ClN2
InChi Key: NQNZNONJZASOKL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CN(CCN1)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)