accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-72-8

IUPAC Name: N-(5-chloro-2,4-dimethoxyphenyl)-3-hydroxynaphthalene-2-carboxamide
Synonyms: N-(5-chloro-2,4-dimethoxyphenyl)-3-hydroxynaphthalene-2-carboxamide
Molecular Weight (g/mol): 357.79
Molecular Formula: C19H16ClNO4
InChi Key: XDWATWCCUTYUDE-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(NC(=O)C2=C(O)C=C3C=CC=CC3=C2)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results