accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-77-3

IUPAC Name: 3-hydroxy-N-phenylnaphthalene-2-carboxamide
Synonyms: naphthol AS
Molecular Weight (g/mol): 263.3
Molecular Formula: C17H13NO2
InChi Key: JFGQHAHJWJBOPD-UHFFFAOYSA-N
SMILES: OC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results