accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 92-81-9

IUPAC Name: 9,10-dihydroacridine
Synonyms: acridan
Molecular Weight (g/mol): 181.24
Molecular Formula: C13H11N
InChi Key: HJCUTNIGJHJGCF-UHFFFAOYSA-N
SMILES: C1C2=CC=CC=C2NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results