accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 92-93-3

IUPAC Name: 4-nitro-1,1'-biphenyl
Synonyms: 4-nitrobiphenyl P-nitrobiphenyl
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: BAJQRLZAPXASRD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results