CAS RN 92-95-5
IUPAC Name:
4-isocyanato-1,1'-biphenyl
Synonyms:
p-xenylcarbimide
Molecular Weight (g/mol):
195.22
Molecular Formula:
C13H9NO
InChi Key:
WIRPZDICFIIBRF-UHFFFAOYSA-N
SMILES:
O=C=NC1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich 4-Biphenylyl isocyanate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.