accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO2H2H

CAS RN 920-42-3

IUPAC Name: (²H)methan(²H)oic acid
Synonyms: (²H)methan(²H)oic acid
Molecular Weight (g/mol): 48.04
Molecular Formula: CH2O2
InChi Key: BDAGIHXWWSANSR-PFUFQJKNSA-N
SMILES: [2H]OC([2H])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results