accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 920304-57-0

IUPAC Name: N,N-diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: N,N-diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 275.2
Molecular Formula: C16H26BNO2
InChi Key: WVPMYKCIKQQJIV-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results