accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOONHClClClOOCH3OOCH3OOH3C

CAS RN 92052-29-4

IUPAC Name: [3,4,5-tris(acetyloxy)-6-[(2,2,2-trichloroethanimidoyl)oxy]oxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-[(2,2,2-trichloroethanimidoyl)oxy]oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 492.68
Molecular Formula: C16H20Cl3NO10
InChi Key: IBUZGVQIKARDAF-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(=N)C(Cl)(Cl)Cl)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results