accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNOH

CAS RN 920966-16-1

IUPAC Name: tert-butyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 229.32
Molecular Formula: C12H23NO3
InChi Key: LBJSEPNOVVUVJA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCCC(CO)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)