accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 920966-30-9

NH2NHCl

CAS RN 920966-30-9

IUPAC Name: 3-aminocyclohexane-1-carbonitrile hydrochloride
Synonyms: 3-aminocyclohexane-1-carbonitrile hydrochloride
Molecular Weight (g/mol): 160.65
Molecular Formula: C7H13ClN2
InChi Key: ZBUXGIYHCTWUTA-UHFFFAOYNA-N
SMILES: Cl.NC1CCCC(C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results