accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 921938-48-9

IUPAC Name: 2,3,4,5,6-pentafluorophenyl pyrimidine-5-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl pyrimidine-5-carboxylate
Molecular Weight (g/mol): 290.15
Molecular Formula: C11H3F5N2O2
InChi Key: TZPLYNSLIVPKAH-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=CN=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results