accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 921938-61-6

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 3-(pyridin-2-yloxy)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 3-(pyridin-2-yloxy)benzoate
Molecular Weight (g/mol): 381.26
Molecular Formula: C18H8F5NO3
InChi Key: LKCOLTBNDUOLEF-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC(OC3=CC=CC=N3)=CC=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results