accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OOOCH3

CAS RN 921938-75-2

IUPAC Name: methyl 3-[3-(dimethylamino)propoxy]benzoate
Synonyms: methyl 3-[3-(dimethylamino)propoxy]benzoate
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: WGAZXKVAWOAUOI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OCCCN(C)C)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results