accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 921938-81-0

IUPAC Name: {4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-7-yl}methanol
Synonyms: {4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazin-7-yl}methanol
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: PPBFQXGUORHMBA-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1N=CC(CO)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results