accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 921938-83-2

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazine-7-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazine-7-carboxylate
Molecular Weight (g/mol): 360.24
Molecular Formula: C15H9F5N2O3
InChi Key: WEFDLXWSZQWZNH-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1N=CC(=C2)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results