accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 921938-96-7

IUPAC Name: 4-[(6-methylpyrazin-2-yl)oxy]benzoyl chloride
Synonyms: 4-[(6-methylpyrazin-2-yl)oxy]benzoyl chloride
Molecular Weight (g/mol): 248.67
Molecular Formula: C12H9ClN2O2
InChi Key: HGYASQNSEYDOEQ-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=C(C=C2)C(Cl)=O)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results