accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNFFFOClO

CAS RN 921939-09-5

IUPAC Name: 1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride
Synonyms: 1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carbonyl chloride
Molecular Weight (g/mol): 304.65
Molecular Formula: C12H8ClF3N2O2
InChi Key: SNBCZQCICOQZPC-UHFFFAOYSA-N
SMILES: CN1N=C(C(C(Cl)=O)=C1OC1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results