accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 921939-14-2

IUPAC Name: methyl({[2-(thiophen-2-yl)pyrimidin-5-yl]methyl})amine
Synonyms: methyl({[2-(thiophen-2-yl)pyrimidin-5-yl]methyl})amine
Molecular Weight (g/mol): 205.28
Molecular Formula: C10H11N3S
InChi Key: VZHVJAYQGDCZAB-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(N=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results