accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2Cl

CAS RN 922-15-6

IUPAC Name: methyl 2-(chloromethyl)prop-2-enoate
Synonyms: methyl 2-(chloromethyl)prop-2-enoate
Molecular Weight (g/mol): 134.56
Molecular Formula: C5H7ClO2
InChi Key: NYMDTEIPYQNXIL-UHFFFAOYSA-N
SMILES: COC(=O)C(=C)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results