CAS RN 922-61-2
CAS RN 922-61-2
IUPAC Name:
(2E)-3-methylpent-2-ene
Synonyms:
trans-3-methyl-2-pentene
Molecular Weight (g/mol):
84.16
Molecular Formula:
C6H12
InChi Key:
BEQGRRJLJLVQAQ-GQCTYLIASA-N
SMILES:
CC\C(C)=C\C
1
–
1
of
1
results
1
3-Methyl-2-pentene (cis- and trans- mixture) 99.0+%, TCI America™
CAS: 922-61-2 Molecular Formula: C6H12 Molecular Weight (g/mol): 84.162 MDL Number: MFCD00009334 InChI Key: BEQGRRJLJLVQAQ-XQRVVYSFSA-N PubChem CID: 643935 IUPAC Name: (Z)-3-methylpent-2-ene SMILES: CCC(=CC)C