accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 922706-40-9

IUPAC Name: 2-(9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 292.19
Molecular Formula: C19H21BO2
InChi Key: WVJQQLZHOADEAF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=CC=CC=C1C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results