accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92273-73-9

IUPAC Name: bromozincylium; butan-1-ide
Synonyms: bromozincylium; butan-1-ide
Molecular Weight (g/mol): 202.4
Molecular Formula: C4H9BrZn
InChi Key: PRWJKDARXFAPOY-UHFFFAOYSA-M
SMILES: [Zn+]Br.CCC[CH2-]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)