accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SSNH2HOOOHO

CAS RN 923-32-0

IUPAC Name: 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid
Synonyms: cystine cistina α-diamino-β-dithiolactic acid cystine DL-form L-(-)-cystine dl-β,β'-dithioalanine cystin zystin β,β'-dithioalanine, dl-
Molecular Weight (g/mol): 240.29
Molecular Formula: C6H12N2O4S2
InChi Key: LEVWYRKDKASIDU-UHFFFAOYNA-N
SMILES: NC(CSSCC(N)C(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)