accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFHOFFHO

CAS RN 92339-07-6

IUPAC Name: [2,3,5,6-tetrafluoro-4-(hydroxymethyl)phenyl]methanol
Synonyms: [2,3,5,6-tetrafluoro-4-(hydroxymethyl)phenyl]methanol
Molecular Weight (g/mol): 210.13
Molecular Formula: C8H6F4O2
InChi Key: SDHKGYDQOGCLQM-UHFFFAOYSA-N
SMILES: OCC1=C(F)C(F)=C(CO)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results