accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92367-11-8

IUPAC Name: 3-[4-(bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
Synonyms: 3-[4-(bromomethyl)phenyl]-3-(trifluoromethyl)diazirine
Molecular Weight (g/mol): 279.06
Molecular Formula: C9H6BrF3N2
InChi Key: BCFUQXOZHPIAPS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1(N=N1)C1=CC=C(CBr)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results