accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 924-42-5

IUPAC Name: N-(hydroxymethyl)prop-2-enamide
Synonyms: N-(hydroxymethyl)acrylamide
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: CNCOEDDPFOAUMB-UHFFFAOYSA-N
SMILES: OCNC(=O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results