accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOPOOFFF

CAS RN 92466-70-1

IUPAC Name: oxobis(2,2,2-trifluoroethoxy)-λ⁵-phosphanylium
Synonyms: oxobis(2,2,2-trifluoroethoxy)-λ⁵-phosphanylium
Molecular Weight (g/mol): 245.04
Molecular Formula: C4H4F6O3P
InChi Key: IMDCVAFSSZPRRM-UHFFFAOYSA-N
SMILES: FC(F)(F)CO[P+](=O)OCC(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)