accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 925-47-3

IUPAC Name: ethyl 2-[(2-ethoxy-2-oxoethyl)sulfanyl]acetate
Synonyms: ethyl 2-[(2-ethoxy-2-oxoethyl)sulfanyl]acetate
Molecular Weight (g/mol): 206.26
Molecular Formula: C8H14O4S
InChi Key: TVCSSJHLVLMADJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CSCC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results