accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 925-60-0

IUPAC Name: propyl prop-2-enoate
Synonyms: 2-propenoic acid, propyl ester
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: PNXMTCDJUBJHQJ-UHFFFAOYSA-N
SMILES: CCCOC(=O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results