accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 925-83-7

IUPAC Name: decanedihydrazide
Synonyms: decanedioic acid, dihydrazide
Molecular Weight (g/mol): 230.31
Molecular Formula: C10H22N4O2
InChi Key: ZWLIYXJBOIDXLL-UHFFFAOYSA-N
SMILES: NNC(=O)CCCCCCCCC(=O)NN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results