accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926-39-6

IUPAC Name: (2-aminoethoxy)sulfonic acid
Synonyms: ethanolamine O-sulfate ethanolamine-O-sulfate
Molecular Weight (g/mol): 141.14
Molecular Formula: C2H7NO4S
InChi Key: WSYUEVRAMDSJKL-UHFFFAOYSA-N
SMILES: NCCOS(O)(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)