accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926-62-5

IUPAC Name: bromo(2-methylpropyl)magnesium
Synonyms: bromo(2-methylpropyl)magnesium
Molecular Weight (g/mol): 161.33
Molecular Formula: C4H9BrMg
InChi Key: CMWBEISSZHZIMU-UHFFFAOYSA-M
SMILES: CC(C)C[Mg]Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)