accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926-64-7

IUPAC Name: 2-(dimethylamino)acetonitrile
Synonyms: acetonitrile, (dimethylamino)
Molecular Weight (g/mol): 84.12
Molecular Formula: C4H8N2
InChi Key: PLXBWEPPAAQASG-UHFFFAOYSA-N
SMILES: CN(C)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results