accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOONOCH3BrOBr

CAS RN 926643-78-9

IUPAC Name: 2,9-dibromo-6,13-dioctyl-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Synonyms: 2,9-dibromo-6,13-dioctyl-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Molecular Weight (g/mol): 648.44
Molecular Formula: C30H36Br2N2O4
InChi Key: HFLBFDNZMQEBCD-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C(=O)C2=CC(Br)=C3C(=O)N(CCCCCCCC)C(=O)C4=CC(Br)=C(C1=O)C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results