accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSNCO

CAS RN 926921-63-3

IUPAC Name: 4-isocyanato-4-(thiophen-2-yl)oxane
Synonyms: 4-isocyanato-4-(thiophen-2-yl)oxane
Molecular Weight (g/mol): 209.26
Molecular Formula: C10H11NO2S
InChi Key: CDJWJYYYKMFRFC-UHFFFAOYSA-N
SMILES: O=C=NC1(CCOCC1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results