accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926921-66-6

IUPAC Name: 4-[(6-methylpyrazin-2-yl)oxy]benzene-1-sulfonyl chloride
Synonyms: 4-[(6-methylpyrazin-2-yl)oxy]benzenesulfonyl chloride
Molecular Weight (g/mol): 284.71
Molecular Formula: C11H9ClN2O3S
InChi Key: GUJMBSPJUZJPEB-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results