accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926921-76-8

IUPAC Name: [(2-bromofuran-3-yl)methyl](methyl)amine hydrochloride
Synonyms: [(2-bromofuran-3-yl)methyl](methyl)amine hydrochloride
Molecular Weight (g/mol): 226.5
Molecular Formula: C6H9BrClNO
InChi Key: GYLBKUAMHLATCB-UHFFFAOYSA-N
SMILES: Cl.CNCC1=C(Br)OC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results