accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 926921-81-5

IUPAC Name: {1-[(thiophen-2-yl)methyl]piperidin-4-yl}methanol
Synonyms: [1-(thiophen-2-ylmethyl)piperidin-4-yl]methanol
Molecular Weight (g/mol): 211.32
Molecular Formula: C11H17NOS
InChi Key: KAZWOHUGLHRSDG-UHFFFAOYSA-N
SMILES: OCC1CCN(CC2=CC=CS2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results