accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 927-67-3

IUPAC Name: propylthiourea
Synonyms: propylthiourea
Molecular Weight (g/mol): 118.2
Molecular Formula: C4H10N2S
InChi Key: UHGKYJXJYJWDAM-UHFFFAOYSA-N
SMILES: CCCNC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)