accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOHOOO

CAS RN 928-04-1

IUPAC Name: potassium 3-carboxyprop-2-ynoate
Synonyms: potassium acetylenedicarboxylate(1-)
Molecular Weight (g/mol): 152.15
Molecular Formula: C4HKO4
InChi Key: KLLYWRUTRAFSJT-UHFFFAOYSA-M
SMILES: [K+].OC(=O)C#CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results